XtraTuff™ Oil Resistant PVC Gloves, Protective Industrial Products
: PIP
These gloves are oil resistant, staying soft and strong even after exposure to machine oils.
- Gauntlet cuff for added forearm protection
- Rough coated palm provides a great grip
Seamless liner provides excellent tactile sensitivity and is treated to resist germs and odors. Used in Cold water, Marine and Handling oily materials.
: Compliant with FDA Part 177 - Indirect food activities.
- Catalog No:
- 20001-874
- 20001-872
- 20001-870
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Finish:
- Rough coated palm
- Rough coated palm
- Rough coated palm
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
Frequently Bought Together