Histo Plas™ Tissue Processing Capsules, Bio Plas | Avantor
Histo Plas™ Tissue Processing Capsules, Bio Plas
White, 29×6 mm
Catalog # 21150-012
Supplier:   Bio Plas
Fournisseur #:  6020
Histo Plas™ Tissue Processing Capsules, Bio Plas
Catalog # 21150-012
Supplier:  Bio Plas
Histo Plas™ Tissue Processing Capsules, Bio Plas
Histo Plas™ Tissue Processing Capsules, Bio Plas
White, 29×6 mm
Price
$272.27
Log in pour la disponibilité
Pack of 500 Case of 5000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Histo Plas™ disposable tissue processing capsule is made of polypropylene
The open mesh construction affords maximum fluid exchange during processing
and the durable marking surface assures permanent identification. Simply place the tissue in the Capsule and press the cover down to lock securely.
The one piece unit is hinged, perfectly aligned and easy to use.
Spécifications
  • Color:
    White
  • Diameter ×H:
    29×6 mm