Ammonium sulfate solution saturée
1 L
Catalog # CA77443-504
New Product
Supplier: Teknova
: A2030
Ammonium sulfate solution saturée
Catalog # CA77443-504
Supplier: Teknova
New Product
CAS Number: 7783-20-2
Jump to:
Product Details & Documents
Sterile saturated ammonium sulfate solution can be used for selective precipitation of proteins..
Synonymes: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
- CAS Number:7783-20-2
- Formule:(NH₄)₂SO₄
- Numéro MDL:MFCD00003391
- Molecular Weight:132.14 g/mol
Spécifications
Résultats des tests de spécification
- Appearance:Liquid
- Solubility:Soluble in water
Spécifications
- Taille:1 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



