VWR® TraceableLIVE® Wi-Fi Data Logging Barometer with Remote Notification
RH: 0 - 100%, non-condensing
P: 500 - 100 mbar, T: 0.01 °C
RH: 0.1%
P: 1 mbar, T: ±0.4 °C
RH: ±3% (5 - 75%), otherwise ±5%
P: ±4 mbar
Product Details & Documents
VWR® TraceableLIVE® is the simple, efficient, and reliable way to stay connected to critical environments, wherever you go
- Monitor barometric pressure, temperature, and humidity readings locally and remotely by connecting to existing Wi-Fi
- Alerts: barometric pressure alarm, humidity alarm, temperature alarm, connectivity interruption, low battery
- Cloud-based data interface with no local software required; web, Android, Apple IOS data visibility
- Continuous data transmission and monitoring using SSL encrypted SECURE connection
- Receive Mobile Push, Email, or Text notifications
- Unlimited scalability; use TraceableLIVE® in individual labs or an entire network of facilities
- Ambient probe and cable probe units available
- Multiple point calibration including barometric pressure, temperature, and humidity; traceable to NIST
The digital datalogging barometer can measure and monitor temperature, humidity, along with barometric pressure. Default WiFi transmission frequency is 15 minutes. Stores up to 100 alarms. Four AAA alkaline battery.
With TraceableLIVE®, stay connected to the lab with a smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser, users can view current humidity and temperatures, control alarm parameters, view datalogging history, and view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on vwr.com (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).
The ideal unit for plants, labs, and all other environments with a need to monitor conditions of environmental air parameters. LCD continuously and simultaneously shows barometric pressure, temperature, and relative humidity. The temperature and humidity sampling rate is 9 seconds, and the barometric pressure sample rate is five minutes and pressure range is 500 to 1100 mb.
Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy. Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement.
Ordering information: Units are supplied with probe, quick start guide, batteries, hook and loop tape, wall mount, Velcro®, magnetic strips, and an ISO 17025 Traceable® certificate.
Spécifications
- Résolution:T: 0.01 °C
RH: 0.1%
P: 1 mbar - Accuracy:T: ±0.4 °C
RH: ±3% (5 - 75%), otherwise ±5%
P: ±4 mbar - Range:T: 0…+55 °C (32…+131 °F)
RH: 0 - 100%, non-condensing
P: 500 - 100 mbar - Cat. No.:76047-266
- W×D×H:108×70×19 mm (4.25×2.75×0.75")
- Interface:WiFi
- Weight:113 g
- Sensor:Ambient