Screw Caps, Firefly | Avantor
Screw Caps, Firefly
Supplier:   FIREFLYSCI, INC.
Screw Caps, Firefly
Supplier:  FIREFLYSCI, INC.
Certificates
Screw Caps, Firefly
Screw Caps, Firefly
2 Options Available Below
Product Details & Documents
Caps are made of polypropylene and are equipped with a poly-vinyl liner.
Ideal for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Availability
    Price
  • Closed Screw Cap with PTFE Septa, 13-425
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.41
    $39.41
    Each
       $39.41
    Supplier Number: SC1
  • Open Screw Cap with PTFE Septa, 13-425
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.41
    $39.41
    Each
       $39.41
    Supplier Number: SC2