Thread Adapters, Polypropylene
Thread Adapters, Polypropylene
Supplier:   BURKLE INC
Thread Adapters, Polypropylene
Supplier:  BURKLE INC
Certificates
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene
23 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Product Family Options
External thread - Internal thread
Product Information
  • Désignation
    Color
    Includes
    Availability
    Price
  • External thread: S 70×6, Internal thread: G 3/4"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $26.24
    $26.24
    Each
       $26.24
    Supplier Number: 0600-0130
    Specifications:
    New Product
    New Product
  • External thread: Mauser, Internal thread: Tri-Sure
    Purple
    Log in for availability
    Log in for availability
    Each
    Each - $76.34
    $76.34
    Each
       $76.34
    Supplier Number: 0600-1012
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Internal thread - Internal thread
Product Information
  • Désignation
    Color
    Includes
    Availability
    Price
  • External thread: Tri-Sure, Internal thread: 2" BSP
    Orange
    Log in for availability
    Log in for availability
    Each
    Each - $76.34
    $76.34
    Each
       $76.34
    Supplier Number: 0600-1006
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: Mauser, Internal thread: 2" BSP
    Blue
    Log in for availability
    Log in for availability
    Each
    Each - $74.73
    $74.73
    Each
       $74.73
    Supplier Number: 0600-1010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: Tri-Sure, Internal thread: Mauser
    Red
    Log in for availability
    Log in for availability
    Each
    Each - $76.34
    $76.34
    Each
       $76.34
    Supplier Number: 0600-1011
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: US Drum thread, Internal thread: 2" BSP
    Silver
    Log in for availability
    Log in for availability
    Each
    Each - $76.34
    $76.34
    Each
       $76.34
    Supplier Number: 0600-1007
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 61 mm (2.40"), Internal thread: 2" BSP
    Grey
    Log in for availability
    Log in for availability
    Each
    Each - $76.34
    $76.34
    Each
       $76.34
    Supplier Number: 0600-1008
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 61 mm (2.40"), Internal thread: 3" Coarse
    Natural
    Log in for availability
    Log in for availability
    Each
    Each - $109.27
    $109.27
    Each
       $109.27
    Supplier Number: 0600-1009
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 64 mm (2.52") (BSI), Internal thread: DIN 60
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $84.78
    $84.78
    Each
       $84.78
    Supplier Number: 0600-1014
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 64 mm (2.52") (BSI), Internal thread: DIN 51
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $97.05
    $97.05
    Each
       $97.05
    Supplier Number: 0600-1016
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 64 mm (2.52") (BSI), Internal thread: DIN 61
    Yellow/blue
    Log in for availability
    Log in for availability
    Each
    Each - $97.05
    $97.05
    Each
       $97.05
    Supplier Number: 0600-1013
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 64 mm (2.52") (BSI), Internal thread: DIN 71
    Brown/blue
    Log in for availability
    Log in for availability
    Each
    Each - $97.05
    $97.05
    Each
       $97.05
    Supplier Number: 0600-1001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Internal thread: 35 mm (1.38") - 2" BSP
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $155.50
    $155.50
    Each
       $155.50
    Supplier Number: 0600-2004
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Internal thread: 51 mm (2.01") - 2" BSP
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $91.53
    $91.53
    Each
       $91.53
    Supplier Number: 0600-2005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Internal thread: DIN 61 - 2" BSP
    Yellow
    Log in for availability
    Log in for availability
    Each
    Each - $76.34
    $76.34
    Each
       $76.34
    Supplier Number: 0600-2001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Internal thread: 63 mm (2.48") - 2" BSP
    White
    Log in for availability
    Log in for availability
    Each
    Each - $91.53
    $91.53
    Each
       $91.53
    Supplier Number: 0600-2006
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Internal thread: DIN 71 - 2" BSP
    Brown
    Log in for availability
    Log in for availability
    Each
    Each - $77.81
    $77.81
    Each
       $77.81
    Supplier Number: 0600-1003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
External thread - External thread
Product Information
  • Désignation
    Color
    Includes
    Availability
    Price
  • External thread: 61 mm (2.40") - Tri-Sure
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $44.05
    $44.05
    Each
       $44.05
    Supplier Number: 0600-3006
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 61 mm (2.40") - 2" BSP
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $44.05
    $44.05
    Each
       $44.05
    Supplier Number: 0600-3007
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 2" coarse Mauser - 64 mm (2.52") (BSI)
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $58.00
    $58.00
    Each
       $58.00
    Supplier Number: 0600-3004
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 2" coarse Tri-Sure - 64 mm (2.52") (BSI)
    Brown
    Log in for availability
    Log in for availability
    Each
    Each - $58.00
    $58.00
    Each
       $58.00
    Supplier Number: 0600-3002
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • External thread: 2" BSP - 64 mm (2.52") (BSI)
    Blue
    Log in for availability
    Log in for availability
    Each
    Each - $58.00
    $58.00
    Each
       $58.00
    Supplier Number: 0600-3003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Adapter set
Product Information
  • Désignation
    Color
    Includes
    Availability
    Price
  • Adapter set thread adapter PP, Internal thread: G 2"
    76077-124, 76077-126, 76077-142, 76077-146
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $249.22
    $249.22
    Pack of 4
       $249.22
    Supplier Number: 0600-3010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.