Aluminum Seals, Tear-Out, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
These tear-out seals are designed to secure septa and stoppers to bottles and vials.
- Tamper-evident center tear-out feature protects target area
- For use with 13 and 20 mm serum finished bottles and vials
- Allow for easy access to septa
Frequently Bought Together