Bacillus cereus Selective Supplement, EMD Millipore
CA1.09875.0010
:
- Pack type:Vial
- Description:Bacillus cereus selective supplement
- Size:10 x 1
- Cat. no.:CA1.09875.0010
- Supplier No.:1.09875.0010
This Bacillus cereus Selective Supplement contains polymyxin B sulfate in lyophilized form.