ContactUsLinkComponent
Automatic Saybolt and ASTM Colorimeter, Koehler Instrument
Catalog # 10118-968
Supplier:  Koehler
Automatic Saybolt and ASTM Colorimeter, Koehler Instrument
Catalog # 10118-968
Supplier:  Koehler
Supplier Number:  K13150
Specifications
  • Range:
    410 - 710 nm
  • Description:
    Automatic Saybolt and ASTM Colorimeter
  • Height:
    17 cm
  • Length:
    19.5 cm
  • Power requirements:
    115 – 240 V, 50/60 Hz
  • Reproducibility:
    ±0.25% T, ±1Saybolt value
  • Cat. no.:
    10118-968
  • Weight:
    7.75 kg
  • Width:
    51.5 cm
  • Supplier No.:
    K13150
Specifications
About this item
This high precision spectrophotometer for objective color analysis of petroleum fuels, oils, waxes and petrochemicals according to the Saybolt and ASTM color scales
  • Test results can also be displayed in terms of CIE values and spectral data
  • Routine diagnostic test allows users to conduct periodic checks on the instrument or to identify faults
  • Direct access of the precision filament lamp from outside the instrument allows for easy replacement
  • Equipped with integrated heater unit for melting solid samples such as fats and waxes and preventing from solidification within the cell during testing
The colorimeter is rugged with a fabricated steel housing which is designed to function equally as a QC instrument within the laboratory or on 24 hour operation in a production environment
Certifications: Conforms to ASTM D156, D1500, D6045, and related test specifications.
Delivery: The colorimeter is supplied with a colored glass filter of known Saybolt value for regular conformance testing.