ContactUsLinkComponent
Pierce™ Coomassie Plus™ (Bradford) Protein Assay Kit, Thermo Scientific
Catalog # CAPI23236
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ Coomassie Plus™ (Bradford) Protein Assay Kit, Thermo Scientific
Catalog # CAPI23236
Supplier:  Thermo Scientific
Supplier Number:  23236
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Coomassie Plus™ – The Better Bradford Assay Kit
  • Size:
    630 Tube Assays/3160 Microplate Assays
  • Cat. no.:
    CAPI23236
  • Detectable Concentration:
    1 - 2000 µg/ml
  • Supplier No.:
    23236
Specifications
About this item
The Pierce™ Coomassie Plus™ Protein Assay is a quick and ready-to-use, reducing agent-compatible, improved Bradford assay to quickly measure (A595 nm) total protein concentration compared to a protein standard.
  • Colorimetric measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use, only single reagent; no working reagent preparation required
  • Almost immediate color development; add, mix, and read results
  • Detects protein concentration in the range 1 to 1,500 µg/mL
  • Improved linearity and response-uniformity compared to traditional Bradford formulations
  • Microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, the Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
Delivery: Kit contains Pierce™ Coomassie Plus™ Protein Assay Reagent (950 mL); Albumin Standard Ampoules (2 mg/mL, 10x1 mL).