Headspace Vials and Caps, MicroSolv
À propos de cet article
MicroSolv brand crimp top and screw top headspace vials in clean packs. Agilent, CTC, PerkinElmer and Varian compatible.
- Meet all instrument OEM specs
- SPME style available
These vials are compatible to most instruments on the market.
: Vials are QC tested.
: Supplied in special boxes with lids to keep the vials clean.
33 Options Available Below
Product Details & Documents
MicroSolv brand crimp top and screw top headspace vials in clean packs. Agilent, CTC, PerkinElmer and Varian compatible.
- Meet all instrument OEM specs
- SPME style available
These vials are compatible to most instruments on the market.
: Vials are QC tested.
: Supplied in special boxes with lids to keep the vials clean.
Product Family Options
Headspace Vials
Informations sur le produit
- DésignationCapacitéMaximum CapacityBottom StyleDiameter ×HMatièreColorAvec bouchonSterilityAvailabilityPrice
- MicroSolv™ screw top vial20.00 ml20.00 mlRounded bottom22.5×75.5 mmBorosilicate glassAmberNonNon sterileLog in pour la disponibilitéPack of 100$346.08Supplier Number: 95020-0AV
Headspace Vials, CTC Compatible
Informations sur le produit
- DésignationCapacitéMaximum CapacityBottom StyleDiameter ×HMatièreColorAvec bouchonSterilityAvailabilityPrice
- MicroSolv™ crimp top vial10.00 ml10.00 mlRounded bottom (HS bottom)22.5×46mmGlassAmberNonNon sterileLog in pour la disponibilitéPack of 100$179.95Supplier Number: 95025-CT-10A
- MicroSolv™ crimp top vial10.00 ml10.00 mlRounded bottom (HS bottom)22.5×46mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$134.66Supplier Number: 95025-CT-10C
- MicroSolv™ crimp top vial20.00 ml20.00 mlRounded bottom (HS bottom)22.5×75.5mmGlassAmberNonNon sterileLog in pour la disponibilitéPack of 100$233.91Supplier Number: 95025-CT-20A
- MicroSolv™ crimp top vial20.00 ml20.00 mlRounded bottom (HS bottom)22.5×75.5mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$182.54Supplier Number: 95025-CT-20C
- MicroSolv™ crimp top vial20.00 ml20.00 mlFlat bottom22.5×75.5mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$194.39Supplier Number: 95025-SPME
- MicroSolv™ screw top vial20.00 ml20.00 mlRounded bottom (HS bottom)22.5×75.5mmGlassAmberNonNon sterileLog in pour la disponibilitéCase of 1,000$3,146.23Supplier Number: 95020-0AV-K
- MicroSolv™ screw top vial20.00 ml20.00 mlRounded bottom (HS bottom)22.5×46mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$200.51Supplier Number: 95020-0CV
- MicroSolv™ screw top vial20.00 ml20.00 mlRounded bottom (HS bottom)22.5×75.5mmGlassClearNonNon sterileLog in pour la disponibilitéCase of 1,000$1,637.46Supplier Number: 95020-0CV-K
- MicroSolv™ screw top vial10.00 ml10.00 mlRounded bottom (HS bottom)22.5×46mmGlassAmberNonNon sterileLog in pour la disponibilitéPack of 100$275.06Supplier Number: 95010-0AV
- MicroSolv™ screw top vial10.00 ml10.00 mlRounded bottom (HS bottom)22.5×46mmGlassAmberNonNon sterileLog in pour la disponibilitéCase of 1,000$2,177.39Supplier Number: 95010-0AV-K
- MicroSolv™ screw top vial10.00 ml10.00 mlRounded bottom (HS bottom)22.5×46mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$152.66Supplier Number: 95010-0CV
- MicroSolv™ screw top vial10.00 ml10.00 mlRounded bottom (HS bottom)22.5×46mmGlassClearNonNon sterileLog in pour la disponibilitéCase of 1,000$1,390.93Supplier Number: 95010-0CV-K
- BASIK™ screw top vial10.00 ml10.00 mlRounded bottom22.5×46 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéCase of 1,000$1,114.83Supplier Number: 95010-E-0CV-K
- BASIK™ screw top vial20.00 ml20.00 mlRounded bottom22.5×75.5 mmBorosilicate glassAmberNonNon sterileLog in pour la disponibilitéCase of 1,000$1,643.64Supplier Number: 95020-E-0AV-K
- BASIK™ screw top vial20.00 ml20.00 mlRounded bottom22.5×75.5 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéPack of 100$166.29Supplier Number: 95020-E-0CV
- BASIK™ screw top vial20.00 ml20.00 mlRounded bottom22.5×75.5 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéCase of 1,000$1,429.20Supplier Number: 95020-E-0CV-K
- BASIK™ screw top vial10.00 ml10.00 mlRounded bottom22.5×46 mmBorosilicate glassAmberNonNon sterileLog in pour la disponibilitéCase of 1,000$1,500.64Supplier Number: 95010-E-0AV-K
- BASIK™ screw top vial10.00 ml10.00 mlRounded bottom22.5×46 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéPack of 100$125.13Supplier Number: 95010-E-0CV
- BASIK™ screw top vial20.00 ml20.00 mlRounded bottom22.5×75.5 mmBorosilicate glassAmberNonNon sterileLog in pour la disponibilitéPack of 100$175.15Supplier Number: 95020-E-0AV
- BASIK™ screw top vial10.00 ml10.00 mlRounded bottom22.5×46 mmBorosilicate glassAmberNonNon sterileLog in pour la disponibilitéPack of 100$160.84Supplier Number: 95010-E-0AV
Headspace Vials, Agilent™ Compatible
Informations sur le produit
- DésignationCapacitéMaximum CapacityBottom StyleDiameter ×HMatièreColorAvec bouchonSterilityAvailabilityPrice
- BASIK™ crimp top vial6.00 ml6.00 mlFlat bottom22×38 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéPack of 100$122.87Supplier Number: 95025-AG-06C
- MicroSolv™ crimp top vial, long neck10.00 ml10.00 mlFlat bottom22.5×46mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$125.42Supplier Number: 95025-AG-10C
- MicroSolv™ crimp top vial, long neck20.00 ml20.00 mlFlat bottom22.5×75.5mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$151.30Supplier Number: 95025-AG-20C
- BASIK™ TapeRed neck crimp top vial20.00 ml20.00 mlFlat bottom23×75 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéPack of 100$121.11Supplier Number: 95025-AP-20C
- BASIK™ TapeRed neck crimp top vial10.00 ml10.00 mlFlat bottom23×46 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéPack of 100$115.05Supplier Number: 95025-AP-10C
- BASIK™ TapeRed neck crimp top vial6.00 ml6.00 mlFlat bottom22×38 mmBorosilicate glassClearNonNon sterileLog in pour la disponibilitéPack of 100$101.94Supplier Number: 95025-AP-06C
Headspace Vials, PerkinElmer Compatible
Informations sur le produit
- DésignationCapacitéMaximum CapacityBottom StyleDiameter ×HMatièreColorAvec bouchonSterilityAvailabilityPrice
- MicroSolv™ crimp top vial5.00 ml5.00 mlRounded bottom (HS bottom)22×38.2mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$122.62Supplier Number: 95025-PE-05C
- MicroSolv™ crimp top vial20.00 ml20.00 mlRounded bottom (HS bottom)23×75.5mmGlassAmberNonNon sterileLog in pour la disponibilitéPack of 100$232.64Supplier Number: 95025-PE-20A
- MicroSolv™ crimp top vial20.00 ml20.00 mlRounded bottom (HS bottom)23×75.5mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$117.08Supplier Number: 95025-PE-20C
- MicroSolv™ crimp top vial with write-on patch20.00 ml20.00 mlRounded bottom (HS bottom)23×75.5mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$172.01Supplier Number: 95025-PE-20WC
Headspace Vials, Varian Compatible
Informations sur le produit
- DésignationCapacitéMaximum CapacityBottom StyleDiameter ×HMatièreColorAvec bouchonSterilityAvailabilityPrice
- MicroSolv™ crimp top vial5.00 ml5.00 mlFlat bottom20×38mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$136.89Supplier Number: 95025-VN-05C
- MicroSolv™ crimp top vial10.00 ml10.00 mlFlat bottom20×54.5mmGlassClearNonNon sterileLog in pour la disponibilitéPack of 100$157.18Supplier Number: 95025-VN-10C
Headspace Crimp Caps
Informations sur le produit
- Catalog #DésignationMatièreSeptumTailleColorAvailabilityPrice
- Safety Crimp CapAluminumTransparent Blue Silicone/White PTFE 45° Shore A, 3.0 mm20 mm, 10 mm HoleSilverLog in pour la disponibilitéPack of 100$286.04
Headspace Screw Caps
Informations sur le produit
- Catalog #DésignationMatièreSeptumTailleColorAvailabilityPrice
- Magnetic Screw CapAluminumSilicone White/PTFE Blue, 50° Shore A, 1.5 mm18 mm Precision Threads, 8 mm HoleSilverLog in pour la disponibilitéPack of 100$175.20
Accessories
Informations sur le produit
- Catalog #Seal SizeAvailabilityPrice
- Black Crimper for Aluminum Crimp Caps, with hex adjustment keyBlack Crimper for Aluminum Crimp Caps, with hex adjustment key97052-28011 mmLog in pour la disponibilitéEach - $849.71
Informations sur le produit
- Catalog #Seal SizeAvailabilityPrice
- Black Crimper for Headspace Aluminum Caps, with hex adjustment keyBlack Crimper for Headspace Aluminum Caps, with hex adjustment key97054-98620 mmLog in pour la disponibilitéPack of 1 - $986.27