Isopropyl Alcohol, Non-Sterile, Texwipe®
Supplier: Texwipe
Certificates
About this item
Non-sterile IPA (isopropyl alcohol) is filtered for cleanliness
- Submicron filtered
- Stream delivery or course spray options are available
- Each bottle is lot coded with an expiration date for easy record keeping
Ideal for surface cleaning, wipe downs and for use with any Texwipe cleanroom wiper.
Delivery: It comes in two convenient dispensers trigger spray (0.5 L) and bulk container (3.8 L).
Packaging: Packing: Filled into cleaned containers, and double-bagged for containers with trigger spray.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together