BD™ FecalSwab™ Fecal Collection and Transport System, BD Diagnostics
75833-310
: BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Type:Single
- Description:Regular Flocked Swab
- Cat. no.:75833-310
- Fill Solution:FecalSwab™ Transport and Preservative Medium
- Supplier No.:220258
FecalSwab™ plastic screw-cap tube with conical shaped bottom filled with 2 mL of FecalSwab™ transport and preservative medium with one swab with flocked nylon fiber tip.
FecalSwab is a convenient system for transporting fecal samples in small instrument-ready tubes, saving space and making it easier to transport to the laboratory.