RODAC™ Contact Plates with Snap Lid, TSA with Lecithin, Polysorbate 80, and Penicillinase, BD Diagnostics
Fournisseur: BD
Certificats
À propos de cet article
The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
- Ensures the integrity of the sample
- Prevents the dish from opening unintentionally
- Protects media from accidental cross contamination
- Improves stability during stacking and storage
Spécifications
- Sterility:Sterile