HiFi Taq DNA Ligase
CA103258-566
:
- Description:HiFi Taq DNA Ligase
- Size:50 Reactions
- Cat. no.:CA103258-566
HiFi Taq DNA Ligase is intended for use in molecular diagnostics applications that depend on high fidelity nick ligation
It is not a substitute for T4 DNA ligase and is not suitable for cloning applications or adapter ligation/NGS library preparations.