ContactUsLinkComponent
Frozen-EZ Yeast Transformation II Kit™, Zymo Research
Catalogue # 76020-472
Fournisseur:  Zymo Research
Numéro CAS:  
undefined
Frozen-EZ Yeast Transformation II Kit™, Zymo Research
Catalogue # 76020-472
Fournisseur:  Zymo Research
Supplier Number:  T2001
Numéro CAS:  
Spécifications
  • Description:
    Frozen-EZ Yeast Transformation II Kit
  • Size:
    120 preps
  • Cat. no.:
    76020-472
Spécifications
À propos de cet article
The Frozen-EZ Yeast Transformation II Kit is designed to make yeast transformations and library screening easier and more efficient than currently available methods.
Yeast cells with high transformation efficiencies can be prepared in under 10 minutes.
Simple method for transforming yeast with single or multiple plasmids in less than 1 hour.
No carrier DNA required.
The yeast cells prepared with the Frozen-EZ Yeast Transformation II Kit™ can be used immediately for transformation or can be stored (i.e., ≤ -70°C) for use at a later time. Yeast prepared with this kit can be transformed with both circular and linear DNAs. Also, the Frozen-EZ Yeast Transformation II Kit™ can be used with other fungi including C. albicans, S. pombe, and P. pastoris.