Bench Top Rotating Sample Divider (BTRSD), Rocklabs
Catalog # CA76047-488
Supplier: ROCKLABS (W/T) CA
Specifications
- Description:Bench Top Rotating Sample Divider
- Electrical:220-240V, 60Hz, 1Ph
- Shipping weight:100 kg
- Width:600 mm
- Depth:496 mm
- Shipping dimensions:825W x 730H x 605D mm
- Height:557 mm
- Hopper volume/throughput:5 kg
- Max. size of feed granules:10 mm
- Motor power:0.1kW, single phase
- Weight:85 kg
- Cat. no.:CA76047-488
Specifications
About this item
The Bench Top Rotating Sample Divider is designed to be placed on a work bench, It can also be used in conjunction with a Bench Top Ring Mill.
- A representative split of any proportion from 2 to 50%
- Two representative splits–sample and remainder
- Sealed bearings
Taking a representative split from a large sample is not as simple as many people believe. Rolling on a mat, multiple grabs or riffle splitting do not produce accurate results. A classic study conducted at Bradford University demonstrated that a rotating sample divider provided the best results.
Ordering information: Manufacturer's 2000 operating hours or one year warranty (whichever comes first), covering all parts (excluding wear parts) and labour if used under normal laboratory conditions. Dust extraction not available.