True RMS Bench Digital Multimeter | Avantor
True RMS Bench Digital Multimeter
Benchtop Multimeter, 10 Hz - 1 MHz
Catalog # 470121-628
Small Business Enterprise
Supplier:   B & K Precision
Fournisseur #:  2831E
True RMS Bench Digital Multimeter
Catalog # 470121-628
Supplier:  B & K Precision
Small Business Enterprise
True RMS Bench Digital Multimeter
True RMS Bench Digital Multimeter
Benchtop Multimeter, 10 Hz - 1 MHz
Price
$1,093.29
Log in pour la disponibilité
Each
Product Details & Documents
A versatile and dependable bench-top digital multimeters suitable for applications.
  • Up to 50,000 count display resolution
  • Basic VDC accuracy of 0.02% for the 5491B and 0.03% for the 2831E
  • Dual display to indicate two measurements simultaneously
  • AC + DC True RMS
  • Up to 25 readings per second measurement rate
Enhance your productivity with built-in math functions and USB connectivity, features not found in other bench meters in this price category.
The meters take typical multimeter measurements such as volts, ohms, and amps with great accuracy and stability and were designed for cost conscious users requiring a basic and dependable meter with a broad range of extra features offered at a value price.
Spécifications
  • Désignation:
    Benchtop Multimeter
  • Fréquence:
    10 Hz - 1 MHz
  • Resistance:
    200 Ohms to 20 Mohms
  • Cat. No.:
    470121-628
  • AC/DC current:
    2mA to 20A (AC/DC)
  • AC/DC voltage:
    200mV to 1000V (AC/DC)