PumpMaster Container Pumps, Burkle
PumpMaster Container Pumps, Burkle
Supplier:   BURKLE INC
PumpMaster Container Pumps, Burkle
Supplier:  BURKLE INC
Certificates
PumpMaster Container Pumps, Burkle
PumpMaster Container Pumps, Burkle
3 Options Available Below
Product Details & Documents
The manual pressure pump is ideally suited for drawing off, decanting and emptying canisters and barrels.
  • Complete with three bungs in different sizes
The hand pump is used to create excess pressure that forces the liquid out of the container. The stopcock also allows extraction of small quantities of liquid. The PumpMaster container pump is mounted on the container using a flexible stopper with a variable and adjustable diameter. No special thread adapters are required. PumpMaster fits all commercial container openings with a diameter from 46.5 mm to 60 mm (1.83 to 2.36"). The immersion tube has a length of 95 cm (37.40") and it can be shortened depending on the size of the container.
Packaging: One pump per package.
Product Family Options
Product Information
  • Désignation
    Matière
    Capacité
    Immersion
    Color
    Gaskets
    Availability
    Price
  • PumpMaster for Non-Aggressive Liquids
    PP/PE
    8 L/min. (2.11 gpm)
    95 cm (37.40")
    Red
    EPDM
    Log in for availability
    Log in for availability
    Each
    Each - $661.95
    $661.95
    Each
       $661.95
    Supplier Number: 5202-2000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • PumpMaster for Petrochemical Liquids
    PP/PE
    8 L/min. (2.11 gpm)
    95 cm (37.40")
    Blue Gray
    Nitrile rubber
    Log in for availability
    Log in for availability
    Each
    Each - $661.95
    $661.95
    Each
       $661.95
    Supplier Number: 5202-3000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • PumpMaster for Acids and Chemical Liquids
    PP/PE
    8 L/min. (2.11 gpm)
    95 cm (37.40")
    Green
    FKM
    Log in for availability
    Log in for availability
    Each
    Each - $920.84
    $920.84
    Each
       $920.84
    Supplier Number: 5202-1000
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Discharge Hose with Shut-Off Valve for PumpMaster for Petrochemical Liquids
    Discharge Hose with Shut-Off Valve for PumpMaster for Petrochemical Liquids
    76047-106
    Log in for availability
    Each - $316.74
Product Information
  • Catalog #
    Availability
    Price
  • Discharge Hose with Shut-Off Valve for PumpMaster for Acids and Chemical Liquids
    Discharge Hose with Shut-Off Valve for PumpMaster for Acids and Chemical Liquids
    76047-114
    Log in for availability
    Each - $412.89
Product Information
  • Catalog #
    Availability
    Price
  • Discharge Hose with Shut-Off Valve for PumpMaster for Non-Aggressive Liquids
    Discharge Hose with Shut-Off Valve for PumpMaster for Non-Aggressive Liquids
    76047-110
    Log in for availability
    Each - $323.12