Neodymium Sphere Magnet
Neodymium Sphere Magnet, 0.5" Diameter
Catalog # 470229-352
Supplier: Master Magnetics
Fournisseur #: 5XNS50BX
Jump to:
Product Details & Documents
A round neodymium magnet for exciting magnet inquiry.
- Very high resistance to demagnetization
- High energy for size
- Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. The sphere magnet has a north and south pole, one on each half of the sphere. It has a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).
Spécifications
- Désignation:Neodymium Sphere Magnet, 0.5" Diameter