Stem Explorations, Levers and Pulleys Building Set
Stem Explorations, Levers and Pulleys Building Set
Stem Explorations, Levers and Pulleys Building Set, 130+
Catalog # 470229-234
Supplier:   K'nex Industries
Supplier #:  79319
Stem Explorations, Levers and Pulleys Building Set
Catalog # 470229-234
Supplier:  K'nex Industries
Stem Explorations, Levers and Pulleys Building Set
Stem Explorations, Levers and Pulleys Building Set
Stem Explorations, Levers and Pulleys Building Set, 130+
Liquidation
$19.65
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Kit of 1 Case of 4
Product Details & Documents
Explore STEM concepts while building lever and pulley models!
  • Students will learn about lever classes, pulley systems, resistance, effort, mechanical advantage and more!
Using the materials included in this set, elementary-school aged children will be engaged and energized as they further their knowledge and understanding of science, technology, engineering and math concepts. The set includes 130+ parts for a single child or team of 2 children to build 3 unique lever or pulley models: a Balance, a Wheelbarrow and a Sailboat. Once built, a downloadable booklet guides students through 3 hands-on, inquiry-based experiments on each of the models.
Spécifications
  • Désignation:
    Stem Explorations, Levers and Pulleys Building Set
  • No. of Parts:
    130+