Rotary Pulse Encoders, Nidec Shimpo
Encoders are ideal for use with panel mount tachometers and counters to monitor motor operation.
- Rotary encoders available from 60 to 1000 ppr, with 5000 rpm maximum capability
- Quadrature versions available
- RE series encoders are ideal for usage with SHIMPO’s panel mount tachometers and counters
- Base or face mounting
- ABS housing enclosure
RE1B-60F: 60 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE1B-600F: 600 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE1B-1000F: 1000 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE2B-30F: Dual (90° out of phase) digital pulses for quadrature applications, 30 pulses/rev., maximum speed of 5000 rpm
RE2B-600F: Dual (90° out of phase) digital pulses for quadrature applications, 600 pulses/rev., maximum speed of 5000 rpm
Current: 40 mA (max)
Cable length: 0.5 m
Temperature limit: −10 to +70 °C
Shaft torque: 30.6 g/cm
- Catalog No:
- 75840-670
- 75840-688
- 75840-668
- 75840-646
- 75840-648
- 75840-650
- 75840-672
- Weight:
- 150 g
- 150 g
- 150 g
- 150 g
- 150 g
- 150 g
- 150 g
Frequently Bought Together