Double Buret Clamp, Hach
Double Buret Clamp, 24 W x 13 L cm (9.5 x 5.2")
Catalog # CA11014-656
Supplier: Hach
Supplier #: 32800
Jump to:
Product Details & Documents
The Double Buret Clamp holds two burets separately, without slippage or obscured graduation lines and numbers.
- Holds burets from micro-size to 100 mL
- Built-in meniscus reader
Specifications
- Désignation:Double Buret Clamp
- Taille:24 W x 13 L cm (9.5 x 5.2")