Test Kits, COD, TNT plus®
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
Product Family Options
- DésignationTailleMéthode chimiqueRange
- COD TNTplus™, ULR (digestion required)25 Tubes/packReactor digestion1 - 60 mg/L CODPack of 25$190.67: TNT820
- COD TNTplus™, ULR (digestion required)25 Tubes/packReactor digestion1 - 60 mg/L COD-: -
- COD TNTplus™, LR (EPA, digestion required)25 Tubes/packReactor digestion3 - 150 mg/L CODPack of 25$192.81: TNT821
- COD TNTplus™, ULR (digestion required)150 Tubes/packReactor digestion1 - 60 mg/L COD-: -
- COD TNTplus™, LR (digestion required)150 Tubes/packReactor digestion3 - 150 mg/L COD-: -
- COD TNTplus™, LR (EPA, digestion required)25 Tubes/packReactor digestion3 - 150 mg/L COD-: -
- COD TNTplus™, HR (digestion required)25 Tubes/packReactor digestion20 - 1500 mg/L CODPack of 25$192.81: TNT822
- COD TNTplus™, LR (EPA, digestion required)150 Tubes/packReactor digestion3 - 150 mg/L COD-: -
- COD TNTplus™, HR (EPA, digestion required)25 Tubes/packReactor digestion20 - 1500 mg/L COD-: -
- COD TNTplus™, HR (digestion required)25 Tubes/packReactor digestion25 - 1000 mg/L CODPack of 25$200.01: TNT825
- COD TNTplus™, HR (EPA, digestion required)150 Tubes/packReactor digestion20 - 1500 mg/L COD-: -
- COD TNTplus™, UHR (digestion required)25 tubes/packReactor digestion250 - 15000 mg/L CODPack of 25$190.64: TNT823
- COD TNTplus™, HR, Hg-Free (digestion required)150 Tubes/packReactor digestion25 - 1000 mg/L COD-: -
- COD TNTplus™, UHR (digestion required)25 tubes/packReactor digestion250 - 15000 mg/L COD-: -
- COD TNTplus™, UHR (digestion required)150 Tubes/packReactor digestion250 - 15000 mg/L COD-: -
- -: -