Volumetric Flasks
Supplier: Wards
Jump to:
À propos de cet article
Class A Flasks.
- Blue graduations
- Glass stopper
Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.
6 Options Available Below
Product Details & Documents
Class A Flasks.
- Blue graduations
- Glass stopper
Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.
Product Family Options
Informations sur le produit
- CapacitéStopper No.ToleranceAvailabilityPrice
- Supplier Number: CH0441A
- Supplier Number: CH0441B
- Supplier Number: CH0441C
- Supplier Number: CH0441E
- Supplier Number: CH0441F
- Supplier Number: CH0441G