Crystallizing Dish
Supplier: Wards
Jump to:
À propos de cet article
Ideal for storage and crystallization.
- Borosilicate glass
- Flat bottom
- Easy pour spout
Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.
4 Options Available Below
Product Details & Documents
Ideal for storage and crystallization.
- Borosilicate glass
- Flat bottom
- Easy pour spout
Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.
Product Family Options
Informations sur le produit
- CapacitéØHauteurAvailabilityPrice
- Supplier Number: CH0066D
- Supplier Number: CH0066E
- 300 ml100 mm50 mmDiscontinued-Supplier Number: CH0066A
- Supplier Number: CH0066B
- Supplier Number: CH0066C