Avantor® Prevail Amide, HPLC Columns
À propos de cet article
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
- Stable from highly organic to highly aqueous
- Specialty phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
Polar embedded phase.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
2 Options Available Below
Product Details & Documents
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
- Stable from highly organic to highly aqueous
- Specialty phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
Polar embedded phase.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
Specifications for Product Family
Spécifications
- Catalog No:
- 76402-318
- 75797-738
- 75797-740
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Spécifications
Product Family Options
Informations sur le produit
- Type de colonnePhaseGranulométrieØ int.LongueurPacking MaterialAvailabilityPrice
- AnalytiqueAmide5 µm4.60 mm100 mmSilicaDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.76402-318Environmentally PreferableEnvironmental certification - Supplier Number: 88660Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.75797-738Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Supplier Number: 88665Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.75797-740Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact