Avantor® Hichrom, Presat HPLC Columns
Fournisseur: Avantor
Certificats
À propos de cet article
Saturation columns are placed between the pump and the injector and they saturate the mobile phase with silica, minimizing dissolution of the analytical column when working above pH 7.
Frequently Bought Together