Needle, Sklar
Product Details & Documents
The Sklar® Needle is used in micro neurosurgical procedures.
- Reusable
- Nonsterile
The needle's straight shaft and sharp tip are designed for manipulation of delicate brain tissue.
Spécifications
- Longueur:190 mm (7½")
- Matière:Stainless steel
- Taille:#12
- Curvature:Straight