Elevator, Sklar | Avantor
Elevator, Sklar
Supplier:   Sklar
Elevator, Sklar
Supplier:  Sklar
Certificates
Elevator, Sklar
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Multipurpose instrument for use in micro neurosurgical procedures.
  • Reusable
  • Nonsterile
The elevator is designed to elevate tissue layers to gain clear access to explore the surgical field. The elevator features a straight shaft and comes with either a curved or angled tip.
Product Family Options
Informations sur le produit
  • Longueur
    Curvature
    Availability
    Price
  • 19.1 cm (7½")
    Angled
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $527.99
    $527.99
    Prix promotionnel pour les nouveaux clients
    Each
       $527.99
    Supplier Number: 98-7315
  • 19.1 cm (7½")
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $562.79
    $562.79
    Prix promotionnel pour les nouveaux clients
    Each
       $562.79
    Supplier Number: 98-7316