ContactUsLinkComponent
Jako Microlaryngeal Hook
  75996-136
 :  Sklar
undefined
Jako Microlaryngeal Hook
  75996-136
 :  Sklar
 :  98-2092
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Length:
    21.6 cm (8½")
  • Material:
    Stainless steel
  • Curvature:
    Angled (90°)
  • Cat. no.:
    75996-136
Sklar's® Jako micro larynx right angled hook is an ENT instrument.
  • Angled 90°
  • Solid round handle
  • Reusable and nonsterile
This hook is used in diagnosis, microsurgery procedures, or to remove foreign bodies from the throat.