BCR® Foam Flat Mop Cover, Berkshire
76028-246
- Description:Foam Flat Mop Cover
- Material:Foam
- Cat. no.:76028-246
Abrasion-resistant, polyester-knit cleanroom fabric laminated to absorbent polyurethane foam.
- Abrasion resistant
- Laser cut
- High absorbency
The V Flat Mop Foam Cover is laser cut and cleanroom laundered to minimize fiber and particle release
This mop cover is compatible with a wide range of chemicals, disinfectants, and cleaners. It is autoclavable and compatible with both gamma and ETO sterilization.
The Flat Mop Foam Cover is abrasion-resistant, and constructed of polyester-knit cleanroom fabric which is laminated to absorbent polyurethane foam. The 10 mm thickness provides a cushioned surface for optimum conformance to uneven floors, walls, and ceilings.
: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.