ContactUsLinkComponent
BCR® Cleanroom Flat Mop Wringer, Berkshire
BCR® Cleanroom Flat Mop Wringer, Berkshire
Catalog # 76028-242
Supplier:  Berkshire
undefined
BCR® Cleanroom Flat Mop Wringer, Berkshire
Catalog # 76028-242
Supplier:  Berkshire
Supplier Number:  BCRVSWRINGER1
Specifications
  • Description:
    Flat Mop Wringer with Handle
  • Material:
    Stainless steel
  • Cat. no.:
    76028-242
Specifications
About this item
The ergonomic, 16" cleanroom flat mop wringer is made of high quality stainless steel, and is suitable for the one-step wet cleaning process.
  • Ergonomic handle
  • High quality stainless steel construction
  • ISO Class 3–8
  • Cleanroom Class 1–100,000
Wringer is made of high-quality stainless steel and durable plastic, suitable for the one-step wet cleaning process. Ergonomically designed handle conveniently serves as a rest for the mop handle while the mop is not in use.