BCR® Cleanroom Flat Mop Wringer, Berkshire
Catalog # 76028-242
Supplier: Berkshire
Specifications
- Description:Flat Mop Wringer with Handle
- Material:Stainless steel
- Cat. no.:76028-242
Specifications
About this item
The ergonomic, 16" cleanroom flat mop wringer is made of high quality stainless steel, and is suitable for the one-step wet cleaning process.
- Ergonomic handle
- High quality stainless steel construction
- ISO Class 3–8
- Cleanroom Class 1–100,000
Wringer is made of high-quality stainless steel and durable plastic, suitable for the one-step wet cleaning process. Ergonomically designed handle conveniently serves as a rest for the mop handle while the mop is not in use.