BCR® Cleanroom Flat Mop Cart, Berkshire
Catalogue # 76028-248
Fournisseur: Berkshire
Spécifications
- Color:Red/grey
- Description:Stainless Steel Mop Cart with Buckets
- Material:Durable Plastic
- Cat. no.:76028-248
Spécifications
À propos de cet article
The Cleanroom Flat Mop Cart is made of high quality stainless steel and is fully autoclavable
- High quality stainless steel
- Fully autoclavable
- Locking casters provide stability during use
The cart includes two 20 liter polypropylene buckets, color-coded blue and red to differentiate between clean and dirty solutions.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.