BCR® Stainless Steel Flat Mop Sieve, Berkshire
Catalog # 76027-822
Supplier: Berkshire
Specifications
- Description:Stainless Steel Flat Mop Sieve
- Material:Stainless steel
- Cat. no.:76027-822
Specifications
About this item
The Flat Mop Sieve is made of high quality stainless steel, and is suitable for the one-step wet cleaning process.
- High quality stainless steel
- Fully autoclavable
Use to release excess waste water into a containment bucket. Simply press the mop onto the sieve to release excess cleaning solution, and apply to surface. Sieve is fully autoclavable.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.