ORP Redox Sensor, Pro Series 1002, YSI
Catalogue # 10218-806
Fournisseur: YSI
Spécifications
- Description:ORP (redox) sensor
- Cat. no.:10218-806
- For Use With:Pro Series
- Supplier No.:605102
Spécifications
À propos de cet article
These sensor measures ORP (oxidation reduction potential) or redox, quickly and easily.
- Easy user-replaceable sensor allows for quick sensor replacements
- One-year warranty