About this item
Restek's FluoroPhenyl phase offers chromatographers the ability to run in reversed-phase or HILIC mode for a variety of compounds.
- Capable of both reversed-phase and HILIC separations
- Ideal for increasing sensitivity and selectivity in LC-MS analyses
- Offers increased retention for charged bases
The culmination of 20+ years in LC, Force performance LC columns are long-lasting and reproducible, even under the stress of UHPLC
Restek's FluoroPhenyl phase offers chromatographers the ability to run in reversed-phase or HILIC mode for a variety of compounds. Restek's FluoroPhenyl columns are also amenable to LC-MS because they are extremely reliable and efficient with acidic mobile phases.
Premium quality is backed by the strength of our 100% Pure Satisfaction guarantee. And perhaps most important, Force columns are fully scalable between 3 or 5 µm HPLC and 1.8 µm UHPLC, so you can easily transfer and optimize methods without extensive calculations.
Specifications
- Catalog No:
- 75854-254
- 75854-276
- 75854-234
- 75854-256
- 75854-278
- 75854-250
- 75854-272
- 75854-274
- 75854-252
- 75854-268
- 75854-246
- 75854-270
- 75854-248
- 75854-264
- 75854-266
- 75854-244
- 75854-262
- 75854-240
- 75854-242
- 75854-258
- 75854-236
- 75854-260
- 75854-238
- Carbon load:
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- pH range:
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- 2.0 - 8.0
- Surface area:
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- 300 m2/g
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43