ContactUsLinkComponent
CIP 220® Acid-Based Process and Research Cleaners
CIP 220® Acid-Based Process and Research Cleaners
Catalog # 77531-094
Supplier:  Steris
New Product
CIP 220® Acid-Based Process and Research Cleaners
Catalog # 77531-094
Supplier:  Steris
Supplier Number:  1D22D3
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    1136 L (300 gal.)
  • Cat. no.:
    77531-094
  • Packaging:
    Plastic tote
Specifications
About this item
CIP 220® Acid detergent is a proprietary blend of hydroxyacetic (glycolic) acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This phosphate free low foaming detergent is ideal for use in CIP, COP and manual applications.
  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Globally Availability - phosphate free, and manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
CIP 220® Acid-based process and research cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable cleaner.