Micro Dissecting Scissors, Sklar
 :   Sklar
Micro Dissecting Scissors, Sklar
 :  Sklar
Micro Dissecting Scissors, Sklar
Micro Dissecting Scissors, Sklar
These micro scissors are used for the dissection of tissues, blood vessels and nerves.
  • Reusable
  • Nonsterile
Available with a flat or round handle, these curved scissors will dissect vessels and nerves. In addition to the tips, the sides may be used for dissection with the tips closed, similar to using forceps.
Product Family Options
  • Désignation
    Longueur
    Matière
    Blade Length
    Blade Type
    Curvature
  • Micro Dissecting Scissors, Flat Handle
    108 mm (4¼")
    Stainless steel
    14 mm (⁹⁄₁₆")
    Blunt/Blunt, Smooth
    Curved
    Each
    Each - 2 074,40 $
    2 074,40 $
    Each
       2 074,40 $
     : 98-7807
    Specifications
    Cat. No.
    75996-298
  • Micro Dissecting Scissors, Flat Handle
    152 mm (6")
    Stainless steel
    14 mm (⁹⁄₁₆")
    Blunt/Blunt, Smooth
    Curved
    Each
    Each - 2 459,62 $
    2 459,62 $
    Each
       2 459,62 $
     : 98-7808
    Specifications
    Cat. No.
    75996-300
  • Micro Dissecting Scissors, Round Handle
    152 mm (6")
    Stainless steel
    14 mm (⁹⁄₁₆")
    Blunt/Blunt, Smooth
    Curved
    Each
    Each - 2 459,62 $
    2 459,62 $
    Each
       2 459,62 $
     : 98-7818
    Specifications
    Cat. No.
    75996-310
  • Micro Dissecting Scissors, Flat Handle
    152 mm (6")
    Stainless steel
    19 mm
    Blunt/Blunt, Smooth
    Curved
    Each
    Each - 2 459,62 $
    2 459,62 $
    Each
       2 459,62 $
     : 98-7809
    Specifications
    Cat. No.
    75996-302
  • Micro Dissecting Scissors, Flat Handle
    152 mm (6")
    Stainless steel
    19 mm
    Blunt/Blunt, Saw Edge
    Curved
    Each
    Each - 2 459,62 $
    2 459,62 $
    Each
       2 459,62 $
     : 98-7811
    Specifications
    Cat. No.
    75996-304
  • Micro Dissecting Scissors, Round Handle
    178 mm (7")
    Stainless steel
    14 mm (⁹⁄₁₆")
    Blunt/Blunt, Smooth
    Curved
    Each
    Each - 2 858,17 $
    2 858,17 $
    Each
       2 858,17 $
     : 98-7819
    Specifications
    Cat. No.
    75996-312