Mini Spore Ampoules, Crosstex®
Mini Spore Ampoules, Clear, 0.4 ml, 32×6.4 mm
Supplier: Crosstex
Supplier #: NMOS1-50-06
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Crosstex Mini Spore ampules are self-contained biological indicators for monitoring steam processes consisting of a hermetically sealed glass ampule which contains modified Trypticase® Soy Broth (TSB) with a pH indicator and Geobacillus stearothermophilus with a population level of 106.
- Reduced incubation time of 48 hours
- Visually distinct positive results indicated by the media transitioning from Purple to Yellow
- Saves time by eliminating aseptic culturing required with traditional spore strips
- Negative controls are included in each box
The Mini Spore ampules may be utilized to monitor Steam sterilization efficacy
Documents
Certificates
Search by Lot NumberSpecifications
- Capacité:0.4 ml
- Color:Clear
- Désignation:Mini Spore Ampoules
- Diameter ×H:32×6.4 mm