Column Cutters, PerkinElmer
Column Cutter
Catalog # CA97019-380
Supplier: PerkinElmer
Supplier #: N9301376
Jump to:
Product Details & Documents
This ceramic column cutter is ideal for cutting polyimide fused silica capillary columns and guard columns.
- Easy to hold and simple to use
- All four sides can be used as a cutting tool
Specifications
- Désignation:Column Cutter