ToxinSensor™ Gel Clot Endotoxin Assay Kit, GenScript
Catalogue # 10127-522
Fournisseur: Genscript
Numéro CAS:
Spécifications
- Description:Gel Clot Endotoxin Assay Kit
- Size:1 Kit (40 reactions)
- Cat. no.:10127-522
- Supplier No.:L00351
Spécifications
À propos de cet article
This endotoxin detection system is intended for use as an in vitro end-product endotoxin test for human and animal parenteral drugs, biological products and medical devices.
- Good reproducibility
- Ready-to-use reagents and materials, such as tips, Endotoxin-free tubes
Delivery information: Kit contains LAL Reagent Water 10 mL (4 bottles), Limulus Amebocyte Lysate (LAL) 4 mL (2 vials), E. coli Endotoxin Standard 0.5 EU (2 vials), Endotoxin-free Vials (5 x 16 vials), Endotoxin-free Tips 200 ?L (1 box of 96 Tips), Endotoxin-free Tips 1000 ?L (2 bags of 12 Tips) and Incubation Rack (1 piece).