SHEL LAB Drosophila Refrigerated Peltier Incubator, Sheldon
Fournisseur: Sheldon Manufacturing
Certificats
À propos de cet article
Using innovative peltier cooling technology, drosphila incubators offer a 78% reduction in energy consumption over traditional compressor based refrigerated incubators.
- Robust design
- Whisper-quiet operation
- Over-temperature thermostat and alarm
These incubators are equipped with LED Lighting and Day/Night Controls to create ideal conditions for fruit flies
Independent over-temperature thermostat and alarm.
The robust cabinet design includes a mechanical convection system to provide improved temperature uniformity, digital temperature controller with display, and over temperature limit control. The compressor-free design creates a whisper-quiet operation.
Certifications: TUV, CE, UL, CSA.
Ordering information: Three-year parts and labor warranty (domestic).
Spécifications
- Catalog No:
- 89409-362
- 76458-746
- 76458-748
- Temperature uniformity:
- ±0.5 °C at 20 °C
- ±0.5 °C at 20 °C
- ±0.5 °C at 20 °C
- No. of outlets:
- One internal
- One internal
- One internal