SP Wilmad-LabGlass Siphon ASTM Extraction Apparatus, SP Industries
Fournisseur: SP Industries
Certificats
À propos de cet article
A block-tin condenser fits within a 400 ml flask and this borosilicate glass extraction cup with siphon tube is suspended with wire from the condenser.
- Borosilicate glass
Reference: ASTM D 297 Methods for Rubber Products; ASTM D 305 Test Method for Solvent-Extractable Material in Black Pigments.
Frequently Bought Together