Econo™ Small Utility Scissors, Sklar
These scissors are straight and have one serrated blade and one smooth blade.
- Reusable
- Small
Econo™ Utility Scissors are multipurpose instruments used for cutting non-sterile materials such as dressings, casts, and surgical drapes. They are Sklar's floor-grade instruments and are sold in bulk quantities. This item is sold in boxes of 12.
- Longueur:63.5 mm (2½")
- Matière:Stainless steel
- Blade Type:Smooth, Sharp/Sharp, Straight