LumiLoupe Plus
Supplier: CARSON OPTICAL
Jump to:
À propos de cet article
A loupe with a transparent base to allow ambient light to illuminate the viewing object.
- Allows for viewing of an object's fine detail
- Dual lenses for close-up viewing and less distortion
2 Options Available Below
Product Details & Documents
A loupe with a transparent base to allow ambient light to illuminate the viewing object.
- Allows for viewing of an object's fine detail
- Dual lenses for close-up viewing and less distortion