ScienceLine pH Combination Electrodes, SI Analytics
ScienceLine pH Combination Electrodes, SI Analytics
Supplier:   SI Analytics
ScienceLine pH Combination Electrodes, SI Analytics
Supplier:  SI Analytics
Certificates
ScienceLine pH Combination Electrodes, SI Analytics
ScienceLine pH Combination Electrodes, SI Analytics
6 Options Available Below
Product Details & Documents
pH combination electrodes are available with plug head and no cable or BNC plug and fixed cable of one meter length.
Ordering information: Connection cables for electrodes with plug heads are sold separately, contact your Avantor representative for more information.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 97041-830
  • 97041-860
  • 97041-984
  • 97041-998
  • 97041-828
  • 97041-834
  • 97041-838
  • 97041-840
  • 97041-958
  • 97041-994
  • 97042-018
  • 97041-978
  • 97041-980
  • 97041-982
  • 97042-200
  • 97042-202
  • 97042-408
  • 97042-428
  • 97041-970
  • Shape:
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Sphere, except type L 8280 (Dome)
  • Reference system:
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
Spécifications
Product Family Options
Electrodes without Connection Cable
Informations sur le produit
  • Désignation
    Type
    Matière
    Ø
    Longueur
    Connecteur
    Electrolyte
    Body Material
    Plage de température
    pH Range
    Availability
    Price
  • Electrode with standard taper NS 14.5
    N 65
    pH glass A
    10 mm
    103 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    ‒5...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285100067
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-830
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode with standard taper NS 14.5
    H 6580
    pH glass H
    10 mm
    103 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285102565
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-860
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode with standard taper NS 14.5
    H 65
    pH glass H
    10 mm
    103 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-984
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode with standard taper NS 14.5
    N 6980
    pH glass A
    10 mm
    103 mm
    Plug head, without Cable
    KCl 3 mol/L
    Glass
    ‒5...+100 °C
    0 - 14
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-998
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    N 62
    pH glass A
    12 mm
    120 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    ‒5...+100 °C
    0 - 14
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,674.51
    $1,674.51
    Each
       $1,674.51
    Supplier Number: 285100034
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-828
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 62
    pH glass H
    12 mm
    120 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,849.00
    $1,849.00
    Each
       $1,849.00
    Supplier Number: 285100215
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-834
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    A 7780
    pH glass A
    12 mm
    120 mm
    Plug head, without cable
    Gel
    Glass
    ‒5...+80 °C
    0 - 14
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,498.41
    $2,498.41
    Each
       $2,498.41
    Supplier Number: 285101260
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-838
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    L 8280
    pH glass A
    12 mm
    120 mm
    Plug head, without cable
    Referid®
    Glass
    ‒5...+80 °C
    pH 2 - 12
    Discontinued
    Supplier Number: 285101277
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-840
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    L 32
    pH glass A
    12 mm
    120 mm
    Plug head, without cable
    KCl 3 mol/L
    Plastic
    –5...+50 °C
    1 - 13
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-958
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    N 64
    pH glass A
    12 mm
    170 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    ‒5...+100 °C
    0 - 14
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,987.93
    $1,987.93
    Each
       $1,987.93
    Supplier Number: 285100059
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-978
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 61
    pH glass H
    12 mm
    170 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285100207
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-980
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 64
    pH glass H
    12 mm
    170 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,275.09
    $2,275.09
    Each
       $2,275.09
    Supplier Number: 285100231
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-982
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 63
    pH glass H
    12 mm
    320 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285100223
    Spécifications:
    More
    Specifications
    Cat. No.
    97042-200
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 6380
    pH glass H
    12 mm
    320 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    +10...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285102549
    Spécifications:
    More
    Specifications
    Cat. No.
    97042-202
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 61-500
    pH glass H
    12 mm
    500 mm
    Plug head, without Cable
    KCl 3 mol/L
    Glass
    0...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285092583
    Spécifications:
    More
    Specifications
    Cat. No.
    97042-408
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    H 61-600
    pH glass H
    12 mm
    600 mm
    Plug head, without cable
    KCl 3 mol/L
    Glass
    0...+100 °C
    0 - 14
    Discontinued
    Supplier Number: 285092591
    Spécifications:
    More
    Specifications
    Cat. No.
    97042-428
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)
  • Electrode
    N 6480 eis
    pH glass A
    12 mm
    170 mm
    Plug head, without cable
    L 5014, Ag/AgCl ref.
    Glass
    10...40 °C
    0 - 14
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,325.28
    $2,325.28
    Each
       $2,325.28
    Supplier Number: 285092337
    Spécifications:
    More
    Specifications
    Cat. No.
    97041-970
    Shape
    Sphere, except type L 8280 (Dome)
    Reference system
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)