To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Phosphoprotein Phosphate Estimation Assay Kit
- Cat. no.:CAPI23270
- No. of Assays/Kit:500 tube assays or 1920 microplate assays
- Supplier No.:23270
Pierce™ Phosphoprotein Phosphate Estimation Assay Kit enables classification and identification of proteins as phosphorylated (serine and threonine), as well as semi-quantitative assessment of the phosphorylation level.
- Specifically measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only
- Test tube and 96-well microplate protocols included and require less than 90 minutes to perform
- Semi-quantitative, calculate moles of phosphate per mole of protein using the included phosvitin standard
- Format is adaptable to development of specific assays for pure phosphoproteins when previously characterized standards are available
Applications include identification of proteins as containing phosphoserine or phosphothreonine phosphorylations; estimating the amount of pS and pT phosphorylation; developing specific quantitative assays for well-characterised phosphoproteins of interest.