Rotavapor® R-300 Rotary Evaporator with Controller, BUCHI
: Buchi
The R-300 meets the highest expectations in convenience and versatility
- Intuitive user interaction
- Precise operation control
- 5 L flask capacity
- Ergonomic handle
- Combi-Clip
- Additional glass assemblies and configurations
The R-300 Rotavapor® has an ergonomic handle for easy positioning of the evaporation flask in to and out of the water bath. The I-300 interface is the central control unit for all process parameters of the Rotavapor R-300. This includes rotation speed; lift position; heating, cooling, and vapor temperature and pressure control. Push notifications on mobile devices allows connected users to view the status of multiple R-300s. The patented multipurpose Combi-clip with snap lock-mechanism provides for quick flask connection and detachment with one hand.
The I-300 interface provides a central location to regulate each evaporation parameter.
: CE and CSA compliant.
: I-300 standard or I-300 Pro controller interface included.
: All accessories and connections are included for out of the box usage. Optional 1L bath, safety shield, >1L evaporation flasks, different joint size glassware, valves for in-house cooling and vacuum are available as accessories (sold separately). Additional glass assemblies and configurations are available. Please contact your Avantor representative for more information.
- Catalog No:
- CA10808-516
- 10808-516
- CA10808-512
- CA10808-456
- 10808-512
- 10808-452
- 10808-456
- CA10808-452
- Rotation speed:
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- Weight:
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- 13.0 - 13.5 kg
- Shipping dimensions:
- 700L×605W×590H mm
- 700L×605W×590H mm
- 700L×605W×590H mm
- 700L×605W×590H mm
- 700L×605W×590H mm
- 700L×605W×590H mm
- 700L×605W×590H mm
- 700L×605W×590H mm
- Dimensions:
- 400L×615W×320H mm
- 400L×615W×320H mm
- 400L×615W×320H mm
- 400L×615W×320H mm
- 400L×615W×320H mm
- 400L×615W×320H mm
- 400L×615W×320H mm
- 400L×615W×320H mm
- Temperature range:
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
- 20 to 220 °C (5 L flask bath)
Frequently Bought Together