Slotted Mass Set | Avantor
Slotted Mass Set
Mass Set of 8, Labware, (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 g
Catalog # 470017-100
Supplier:   Wards
Fournisseur #:  PH0037F-SS
Slotted Mass Set
Catalog # 470017-100
Supplier:  Wards
Slotted Mass Set
Slotted Mass Set
Mass Set of 8, Labware, (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 g
Price
$239.85
Log in pour la disponibilité
Each
Product Details & Documents
Premium Stainless Steel Slotted Masses.
  • 8 slotted masses included
  • Metal rack with handle included
This premium slotted mass set has accurate stainless steel machines masses placed on a metal and plastic rack with metal handle. Includes 8 masses.
Spécifications
  • Désignation:
    Mass Set of 8, Labware
  • Includes:
    (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 g